5-Aminopyrazine-2-carboxylic acid 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66207-03
H66207-06
H66207-MD
AAH66207-03EA
251.22
CAD
AAH66207-03
AAH66207-06
AAH66207-MD
5-Aminopyrazine-2-carboxylic acid 95%
5-Aminopyrazine-2-carboxylic acid
Purity: 95%
CAS No.: 40155-43-9
Molecular Formula: C5H5N3O2
Formula Weight: 139.11
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 120673
MDL No.: MFCD07371664
Formula:
C₅H₅N₃O₂ MW: 139.11 g/mol Storage Temperature: Freezer |
MDL Number:
MFCD07371664 CAS Number: 40155-43-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...