7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66188-03
H66188-06
H66188-MD
AAH66188-03EA
102.07
CAD
AAH66188-03
AAH66188-06
AAH66188-MD
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline 95%
7-Methoxy-2,2,4-trimethyl-1,2-dihydroquinoline
Purity: 95%
CAS No.: 1810-74-8
Molecular Formula: C13H17NO
Formula Weight: 203.29
Physical Form: Solid
Formula:
C₁₃H₁₇NO Storage Temperature: Ambient |
MDL Number:
MFCD00627122 CAS Number: 1810-74-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...