6-Bromonicotinonitrile 97%

Supplier: Thermo Scientific Chemicals
Warning

H66177-03 H66177-06
AAH66177-03EA 68.1 CAD
AAH66177-03 AAH66177-06
6-Bromonicotinonitrile 97%
6-Bromonicotinonitrile

Purity: 97% CAS No.: 139585-70-9 Molecular Formula: C6H3BrN2 Formula Weight: 183.01 Physical Form: Solid

Formula: C₆H₃BrN₂
Storage Temperature: Ambient
MDL Number: MFCD00234141
CAS Number: 139585-70-9
UN: 3439
ADR: 6.1,III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR