1-Ethyl-1H-imidazole-2-carbaldehyde 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66143-03
H66143-MD
AAH66143-03EA
164.32
CAD
AAH66143-03
AAH66143-MD
1-Ethyl-1H-imidazole-2-carbaldehyde 95%
1-Ethyl-1H-imidazole-2-carbaldehyde
Purity: 95%
CAS No.: 111851-98-0
Molecular Formula: C6H8N2O
Formula Weight: 124.14
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 3538106
MDL No.: MFCD07380875
Formula:
C₆H₈N₂O Storage Temperature: Refrigerator |
MDL Number:
MFCD07380875 CAS Number: 111851-98-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...