8-Bromo-2-chloro-6-fluoroquinazoline 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66128-03
H66128-MD
AAH66128-03EA
541.94
CAD
AAH66128-03
AAH66128-MD
8-Bromo-2-chloro-6-fluoroquinazoline 95%
8-Bromo-2-chloro-6-fluoroquinazoline
Purity: 95%
CAS No.: 953039-63-9
Molecular Formula: C8H3BrClFN2
Formula Weight: 261.48
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 15583920
MDL No.: MFCD18382554
Formula:
C₈H₃BrClFN₂ Storage Temperature: Refrigerator |
MDL Number:
MFCD18382554 CAS Number: 953039-63-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...