cis-4-Aminocyclohexanol hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66110-03
H66110-MD
AAH66110-03EA
439.24
CAD
AAH66110-03
AAH66110-MD
cis-4-Aminocyclohexanol hydrochloride 95%
cis-4-Aminocyclohexanol hydrochloride
Purity: 95%
CAS No.: 56239-26-0
Molecular Formula: C6H13NO.HCl
Formula Weight: 151.63
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Sensitivity: Moisture Sensitive
Melting Point: 193-200°
Beilstein Reference: 3909295
MDL No.: MFCD07366531
Formula:
C₆H₁₄ClNO Melting Pt: 193…200 °C Storage Temperature: Ambient |
MDL Number:
MFCD07366531 CAS Number: 56239-26-0 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...