(5-Fluoro-2-methyl-1H-indol-3-yl)acetic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H66088-03 H66088-06
AAH66088-03EA 274.92 CAD
AAH66088-03 AAH66088-06
(5-Fluoro-2-methyl-1H-indol-3-yl)acetic acid 97%
(5-Fluoro-2-methyl-1H-indol-3-yl)acetic acid

Purity: 97% CAS No.: 71987-67-2 Molecular Formula: C11H10FNO2 Formula Weight: 207.20 Physical Form: Solid

Formula: C₁₁H₁₀FNO₂
Storage Temperature: Ambient
MDL Number: MFCD02664389
CAS Number: 71987-67-2

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR