4-(Trifluoromethyl)indole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66080-03
H66080-MD
AAH66080-03EA
376.04
CAD
AAH66080-03
AAH66080-MD
4-(Trifluoromethyl)indole 97%
4-(Trifluoromethyl)indole
Purity: 97%
CAS No.: 128562-95-8
Molecular Formula: C9H6F3N
Formula Weight: 185.15
Physical Form: Solid
Formula:
C₉H₆F₃N Storage Temperature: Ambient |
MDL Number:
MFCD09954766 CAS Number: 128562-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...