2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66063-03
H66063-MD
AAH66063-03EA
273.34
CAD
AAH66063-03
AAH66063-MD
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone 97%
2',4'-Dihydroxy-2-(4-hydroxyphenyl)acetophenone
Purity: 97%
CAS No.: 17720-60-4
Molecular Formula: C14H12O4
Formula Weight: 244.25
Physical Form: Solid
Formula:
C₁₄H₁₂O₄ Storage Temperature: Ambient |
MDL Number:
MFCD00498162 CAS Number: 17720-60-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...