6-Fluoroquinoline-2-carboxaldehyde 97%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 6-Fluoroquinoline-2-carbaldehyde

H66052-03 H66052-MD H66052-06
AAH66052-03EA 211.72 CAD
AAH66052-03 AAH66052-MD AAH66052-06
6-Fluoroquinoline-2-carboxaldehyde 97%
6-Fluoroquinoline-2-carboxaldehyde

Purity: 97% CAS No.: 260430-93-1 Molecular Formula: C10H6FNO Formula Weight: 175.16 Storage Temperature: 2°C to 8°C Physical Form: Solid Sensitivity: Moisture Sensitive Melting Point: 113-120° Beilstein Reference: 8479194 MDL No.: MFCD06751849

Formula: C₁₀H₆FNO
MW: 175.162 g/mol
Melting Pt: 113…120 °C
Storage Temperature: Ambient
MDL Number: MFCD06751849
CAS Number: 260430-93-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR