1-Ethyl-1-methylpyrrolidinium bromide 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
1-Ethyl-1-methylpyrrolidin-1-ium bromide
H66050-14
H66050-22
AAH66050-14EA
151.52
CAD
AAH66050-14
AAH66050-22
1-Ethyl-1-methylpyrrolidinium bromide 98%
1-Ethyl-1-methylpyrrolidinium bromide
Purity: 98%
CAS No.: 69227-51-6
Molecular Formula: C7H16BrN
Formula Weight: 194.12
Physical Form: Solid
Formula:
C₇H₁₆BrN MW: 194.12 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03095384 CAS Number: 69227-51-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...