(S)-Tetrahydrofuran-3-amine hydrochloride 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H66034-03
H66034-06
H66034-MD
AAH66034-03EA
103.65
CAD
AAH66034-03
AAH66034-06
AAH66034-MD
(S)-Tetrahydrofuran-3-amine hydrochloride 95%
(S)-Tetrahydrofuran-3-amine hydrochloride
Purity: 95%
CAS No.: 204512-95-8
Molecular Formula: C4H10ClNO
Formula Weight: 123.58
Storage Temperature: -30°C to -10°C
Physical Form: Solid
Beilstein Reference: 14637084
MDL No.: MFCD08445642
Formula:
C₄H₁₀ClNO Storage Temperature: Refrigerator |
MDL Number:
MFCD08445642 CAS Number: 204512-95-8 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...