1-Bromo-3-chloro-2-iodobenzene 97%

Supplier: Thermo Scientific Chemicals
Warning

H66020-03 H66020-06 H66020-MD
AAH66020-03EA 98.12 CAD
AAH66020-03 AAH66020-06 AAH66020-MD
1-Bromo-3-chloro-2-iodobenzene 97%
1-Bromo-3-chloro-2-iodobenzene

Purity: 97% CAS No.: 450412-28-9 Molecular Formula: C6H3BrClI Formula Weight: 317.35 Physical Form: Solid Sensitivity: Light Sensitive Beilstein Reference: 9119447 MDL No.: MFCD09907935

Formula: C₆H₃BrClI
MW: 317.348 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09907935
CAS Number: 450412-28-9

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR