2-(1H-Imidazol-1-yl)acetonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

H66012-03 H66012-MD
AAH66012-03EA 290.72 CAD
AAH66012-03 AAH66012-MD
2-(1H-Imidazol-1-yl)acetonitrile 95%
2-(1H-Imidazol-1-yl)acetonitrile

Purity: 95% CAS No.: 98873-55-3 Molecular Formula: C5H5N3 Formula Weight: 107.12 Storage Temperature: -30°C to -10°C Physical Form: Solid Beilstein Reference: 7473110 UN No.: 3439 MDL No.: MFCD06421433

Formula: C₅H₅N₃
Storage Temperature: Refrigerator
MDL Number: MFCD06421433
CAS Number: 98873-55-3
UN: 3439
ADR: "6.1",III

Order Now

SPECIFICATIONS

Environmentally Preferable

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR