Z-Leu-OSu 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Z-Leu-Osu, N-Benzyloxycarbonyl-L-leucine N-succinimidyl ester
H65546-06
H65546-14
H65546-22
AAH65546-06EA
82.63
CAD
AAH65546-06
AAH65546-14
AAH65546-22
Z-Leu-OSu 95%
Z-Leu-OSu
Purity: 95%
CAS No.: 3397-35-1
Molecular Formula: C18H22N2O6
Formula Weight: 362.38
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 116-120°
Beilstein Reference: 1555663
Optical Rotation: -33°(c=1 in dioxane)
MDL No.: MFCD00042759
Formula:
C₁₈H₂₂N₂O₆ MW: 362.382 g/mol Melting Pt: 116…120 °C Storage Temperature: Ambient |
MDL Number:
MFCD00042759 CAS Number: 3397-35-1 EINECS: 222-256-4 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...