4-Bromo-1H-indazole 97+%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-Bromoindazole
H64527-06
H64527-14
AAH64527-03EA
39.85
CAD
AAH64527-03
AAH64527-06
AAH64527-14
4-Bromo-1H-indazole 97+%
4-Bromo-1H-indazole
Purity: 97+%
CAS No.: 186407-74-9
Molecular Formula: C7H5BrN2
Formula Weight: 197.04
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 164-169°
Beilstein Reference: 9757929
UN No.: 2811
MDL No.: MFCD05664001
Formula:
C₇H₅BrN₂ MW: 197.04 g/mol Melting Pt: 164…169 °C Storage Temperature: Ambient |
MDL Number:
MFCD05664001 CAS Number: 186407-74-9 UN: 2811 ADR: "6.1",III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...