N-tert-Butylglycine hydrochloride 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
2-(tert-Butylamino)acetic acid hydrochloride
H64270-03
H64270-06
H64270-14
AAH64270-03EA
43.61
CAD
AAH64270-03
AAH64270-06
AAH64270-14
N-tert-Butylglycine hydrochloride 97%
N-tert-Butylglycine hydrochloride
Purity: 97%
CAS No.: 6939-23-7
Molecular Formula: C6H14ClNO2
Formula Weight: 167.63
Physical Form: Solid
Appearance: White
Melting Point: 223-224°
Beilstein Reference: 6296616
MDL No.: MFCD03788441
Formula:
C₆H₁₄ClNO₂ MW: 167.633 g/mol Melting Pt: 223…224 °C Storage Temperature: Ambient |
MDL Number:
MFCD03788441 CAS Number: 6939-23-7 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...