2-Bromo-5-(trifluoromethyl)phenol 98%

Supplier: Thermo Scientific Chemicals
Danger

Synonyms: 2-Bromo-5-trifluoromethylphenol

H64009-06 H64009-14
AAH64009-06EA 94.8 CAD
AAH64009-06 AAH64009-14
2-Bromo-5-(trifluoromethyl)phenol 98%
2-Bromo-5-(trifluoromethyl)phenol

Purity: 98% CAS No.: 402-05-1 Molecular Formula: C7H4BrF3O Formula Weight: 241.01 Physical Form: Liquid Appearance: Pale brown UN No.: 3265 MDL No.: MFCD09261190

Formula: C₇H₄BrF₃O
MW: 241.01 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09261190
CAS Number: 402-05-1
UN: 3265
ADR: "8",III

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR