Fmoc-D-Abu-OH 98%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (R)-2-(Fmoc-amino)butyric acid

H63923-03 H63923-06 H63923-14
AAH63923-03EA 54.98 CAD
AAH63923-03 AAH63923-06 AAH63923-14
Fmoc-D-Abu-OH 98%
Fmoc-D-Abu-OH

Purity: 98% CAS No.: 170642-27-0 Molecular Formula: C19H19NO4 Formula Weight: 325.36 Physical Form: Solid

Formula: C₁₉H₁₉NO₄
Storage Temperature: Ambient
MDL Number: MFCD00270336
CAS Number: 170642-27-0

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR