5-Chloro[1,2,4]triazolo[4,3-a]pyridine 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
5-Chloro-[1,2,4]triazolo[4,3-a]pyridine
H63881-03
H63881-06
H63881-MD
AAH63881-03EA
251.22
CAD
AAH63881-03
AAH63881-06
AAH63881-MD
5-Chloro[1,2,4]triazolo[4,3-a]pyridine 97%
5-Chloro[1,2,4]triazolo[4,3-a]pyridine
Purity: 97%
CAS No.: 27187-13-9
Molecular Formula: C6H4ClN3
Formula Weight: 153.57
Physical Form: Solid
Formula:
C₆H₄ClN₃ MW: 153.569 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD11109431 CAS Number: 27187-13-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...