3-(1H-Tetrazol-1-yl)aniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H63829-03 H63829-06 H63829-MD
AAH63829-03EA 224.36 CAD
AAH63829-03 AAH63829-06 AAH63829-MD
3-(1H-Tetrazol-1-yl)aniline 97%
3-(1H-Tetrazol-1-yl)aniline

Purity: 97% CAS No.: 14213-12-8 Molecular Formula: C7H7N5 Formula Weight: 161.16 Physical Form: Solid

Formula: C₇H₇N₅
Storage Temperature: Ambient
MDL Number: MFCD06687920
CAS Number: 14213-12-8

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR