6-Bromo-1H-indole-2-carboxylic acid 97%

Supplier: Thermo Scientific Chemicals
Warning

H63799-03 H63799-06 H63799-MD
AAH63799-03EA 184.86 CAD
AAH63799-03 AAH63799-06 AAH63799-MD
6-Bromo-1H-indole-2-carboxylic acid 97%
6-Bromo-1H-indole-2-carboxylic acid

Purity: 97% CAS No.: 16732-65-3 Molecular Formula: C9H6BrNO2 Formula Weight: 240.05 Storage Temperature: 2°C to 8°C Physical Form: Solid MDL No.: MFCD02664469

Formula: C9H6BrNO2
Storage Temperature: Ambient
MDL Number: MFCD02664469
CAS Number: 16732-65-3

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR