7-Methoxyindole 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63767-03
H63767-06
H63767-14
AAH63767-03EA
74.26
CAD
AAH63767-03
AAH63767-06
AAH63767-14
7-Methoxyindole 97%
7-Methoxyindole
Purity: 97%
CAS No.: 3189-22-8
Molecular Formula: C9H9NO
Formula Weight: 147.17
Flash Point: 113°(235°F)
Physical Form: Solid
Density: 1.126
Refractive Index: 1.612
Boiling Point: 108-110°/0.3mm
MDL No.: MFCD00047203
Formula:
C₉H₉NO Boiling Pt: 108…110 °C/0.3 mm Density: 1.126 Flash Pt: 113 °C(235 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00047203 CAS Number: 3189-22-8 EINECS: 221-690-1 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...