Tris(carbobenzoxy)-L-arginine 95%
Supplier: Thermo Scientific Chemicals
Synonyms:
Nα,Nδ,Nomega-Tris(benzyloxycarbonyl)-L-arginine, Tri-Cbz-L-arginine, Z-Arg(Z)2-OH, Z-Arg(Cbz)₂-OH
H63621-03
H63621-06
AAH63621-03EA
75.84
CAD
AAH63621-03
AAH63621-06
Tris(carbobenzoxy)-L-arginine 95%
Tris(carbobenzoxy)-L-arginine
Purity: 95%
CAS No.: 14611-34-8
Molecular Formula: C30H32N4O8
Formula Weight: 576.6
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 139°
Beilstein Reference: 3079013
Optical Rotation: +17°(c=1 in chloroform)
MDL No.: MFCD00038693
Formula:
C₃₀H₃₂N₄O₈ MW: 576.61 g/mol Melting Pt: 139 °C Storage Temperature: Ambient |
MDL Number:
MFCD00038693 CAS Number: 14611-34-8 EINECS: 238-647-8 |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...