4-Chloro-2-(4-morpholinyl)quinazoline 97%

Supplier: Thermo Scientific Chemicals
Warning

H63458-03 H63458-MD
AAH63458-03EA 475.58 CAD
AAH63458-03 AAH63458-MD
4-Chloro-2-(4-morpholinyl)quinazoline 97%
4-Chloro-2-(4-morpholinyl)quinazoline

Purity: 97% CAS No.: 39216-67-6 Molecular Formula: C12H12ClN3O Formula Weight: 249.7 Physical Form: Solid

Formula: C₁₂H₁₂ClN₃O
Storage Temperature: Ambient
CAS Number: 39216-67-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR