3-(Piperazin-1-ylmethyl)-1H-indole 95%

Supplier: Thermo Scientific Chemicals
Warning

H63402-03 H63402-06 H63402-MD
AAH63402-03EA 319.16 CAD
AAH63402-03 AAH63402-06 AAH63402-MD
3-(Piperazin-1-ylmethyl)-1H-indole 95%
3-(Piperazin-1-ylmethyl)-1H-indole

Purity: 95% CAS No.: 114746-66-6 Molecular Formula: C13H17N3 Formula Weight: 215.29 Physical Form: Solid

Formula: C₁₃H₁₇N₃
Storage Temperature: Ambient
CAS Number: 114746-66-6

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR