2-Amino-3-benzyloxypyrazine 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H63162-03
H63162-MD
AAH63162-03EA
339.7
CAD
AAH63162-03
AAH63162-MD
2-Amino-3-benzyloxypyrazine 96%
2-Amino-3-benzyloxypyrazine
Purity: 97%
CAS No.: 110223-15-9
Molecular Formula: C11H11N3O
Formula Weight: 201.22
Physical Form: Solid
Formula:
C₁₁H₁₁N₃O MW: 201.229 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD09838954 CAS Number: 110223-15-9 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...