z-Tyr-OMe 98%
Supplier: Thermo Scientific Chemicals
H63113-06
H63113-14
H63113-22
AAH63113-06EA
77.74
CAD
AAH63113-06
AAH63113-14
AAH63113-22
z-Tyr-OMe 98%
z-Tyr-OMe
Purity: 98%
CAS No.: 13512-31-7
Molecular Formula: C18H19NO5
Formula Weight: 329.35
Storage Temperature: 2°C to 8°C
Physical Form: Solid
Melting Point: 93-97° dec
Optical Rotation: +48°(c=1 in chloroform)
MDL No.: MFCD00038784
Formula:
C₁₈H₁₉NO₅ Melting Pt: 93…97 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD00038784 CAS Number: 13512-31-7 EINECS: 426-320-4 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...