4-Chlorophenethylamine 97%
Supplier: Thermo Scientific Chemicals
|
Danger
|
Synonyms:
2-(4-Chlorophenyl)ethylamine
H63081-06
H63081-14
H63081-22
AAH63081-06EA
54.35
CAD
AAH63081-06
AAH63081-14
AAH63081-22
4-Chlorophenethylamine 97%
4-Chlorophenethylamine
Purity: 97%
CAS No.: 156-41-2
Molecular Formula: C8H10ClN
Formula Weight: 155.62
Flash Point: 106°(223°F)
Physical Form: Liquid
Density: 1.120
Boiling Point: 60-65°/0.1mm
Beilstein Reference: 508247
MDL No.: MFCD00008191
Formula:
C₈H₁₀ClN Boiling Pt: 60…65 °C/0.1 mm Density: 1.120 Flash Pt: 106 °C(223 °F) Storage Temperature: Ambient |
MDL Number:
MFCD00008191 CAS Number: 156-41-2 EINECS: 205-853-4 UN: 2735 ADR: "8",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...