trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62634-03
H62634-06
AAH62634-03EA
309.68
CAD
AAH62634-03
AAH62634-06
trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid 97%
trans-2-(Benzyloxycarbonylamino)cyclohexanecarboxylic acid
Purity: 97%
CAS No.: 61935-48-6
Molecular Formula: C15H19NO4
Formula Weight: 277.31
Physical Form: Solid
Formula:
C₁₅H₁₉NO₄ Storage Temperature: Ambient |
MDL Number:
MFCD03844616 CAS Number: 61935-48-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...