3-Bromo-2,6-difluoroaniline 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62609-03
H62609-06
AAH62609-03EA
303.36
CAD
AAH62609-03
AAH62609-06
AAH62609-MD
3-Bromo-2,6-difluoroaniline 96%
3-Bromo-2,6-difluoroaniline
Purity: 96%
CAS No.: 1262198-07-1
Molecular Formula: C6H4BrF2N
Formula Weight: 208.00
Physical Form: Solid
UN No.: 2811
Formula:
C₆H₄BrF₂N MW: 208.006 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD22627833 CAS Number: 1262198-07-1 UN: 2811 ADR: 6.1,III |
Learn more
About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...