cis-4-Isopropylcyclohexanecarboxylic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62594-03
H62594-06
H62594-MD
AAH62594-03EA
455.04
CAD
AAH62594-03
AAH62594-06
AAH62594-MD
cis-4-Isopropylcyclohexanecarboxylic acid 97%
cis-4-Isopropylcyclohexanecarboxylic acid
Purity: 97%
CAS No.: 7084-93-7
Molecular Formula: C10H18O2
Formula Weight: 170.24
Physical Form: Solid
Formula:
C₁₀H₁₈O₂ Storage Temperature: Ambient |
CAS Number:
7084-93-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...