trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane 97%

Supplier: Thermo Scientific Chemicals

H62487-03 H62487-06 H62487-14
AAH62487-03EA 124.5 CAD
AAH62487-03 AAH62487-06 AAH62487-14
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane 97%
trans-1-(Boc-amino)-4-(hydroxymethyl)cyclohexane

Purity: 97% CAS No.: 239074-29-4 Molecular Formula: C12H23NO3 Formula Weight: 229.31 Physical Form: Solid

Formula: C₁₂H₂₃NO₃
Storage Temperature: Ambient
MDL Number: MFCD03844605
CAS Number: 239074-29-4

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR