2-(3-Bromo-4-fluorophenyl)acetonitrile 96%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H62260-03
H62260-06
AAH62260-03EA
138.25
CAD
AAH62260-03
AAH62260-06
AAH62260-MD
2-(3-Bromo-4-fluorophenyl)acetonitrile 96%
2-(3-Bromo-4-fluorophenyl)acetonitrile
Purity: 96%
CAS No.: 501420-63-9
Molecular Formula: C8H5BrFN
Formula Weight: 214.03
Storage Temperature: 2°C to 8°C
Physical Form: Solid
UN No.: 3439
MDL No.: MFCD08458154
Formula:
C₈H₅BrFN Storage Temperature: Ambient |
MDL Number:
MFCD08458154 CAS Number: 501420-63-9 UN: 3439 ADR: "6.1",III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...