N,1-Di-Boc-L-histidine dicyclohexylammonium salt 95%
Supplier: Thermo Scientific Chemicals
H62018-03
H62018-06
AAH62018-03EA
98.91
CAD
AAH62018-03
AAH62018-06
N,1-Di-Boc-L-histidine dicyclohexylammonium salt 95%
N,1-Di-Boc-L-histidine dicyclohexylammonium salt
Purity: 95%
CAS No.: 31687-58-8
Molecular Formula: C16H25N3O6.C12H23N
Formula Weight: 536.70
Physical Form: Solid
Appearance: White
Melting Point: 160-165° dec
Formula:
C₁₆H₂₅N₃O₆·C₁₂H₂₃N Melting Pt: 160…165 °C (dec.) Storage Temperature: Ambient |
MDL Number:
MFCD06804675 CAS Number: 31687-58-8 EINECS: 250-764-6 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...