trans-2-(Boc-amino)cyclohexanecarboxylic acid 97%

Supplier: Thermo Scientific Chemicals

H62007-03 H62007-06 H62007-MD
AAH62007-03EA 211.72 CAD
AAH62007-03 AAH62007-06 AAH62007-MD
trans-2-(Boc-amino)cyclohexanecarboxylic acid 97%
trans-2-(Boc-amino)cyclohexanecarboxylic acid

Purity: 97% CAS No.: 209128-50-7 Molecular Formula: C12H21NO4 Formula Weight: 243.29 Physical Form: Solid

Formula: C₁₂H₂₁NO₄
Storage Temperature: Ambient
MDL Number: MFCD02094138
CAS Number: 209128-50-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR