2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile 95%

Supplier: Thermo Scientific Chemicals
Warning

H61858-03 H61858-MD
AAH61858-03EA 458.2 CAD
AAH61858-03 AAH61858-MD
2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile 95%
2-Bromo-6-(4-methyl-1-piperazinyl)benzonitrile

Purity: 95% CAS No.: 1260763-01-6 Molecular Formula: C12H14BrN3 Formula Weight: 280.17 Physical Form: Solid UN No.: 3439

Formula: C₁₂H₁₄BrN₃
MW: 280.169 g/mol
Storage Temperature: Ambient
CAS Number: 1260763-01-6
UN: 3439
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR