2-Amino-3-bromobenzonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61823-03
H61823-MD
AAH61823-03EA
474
CAD
AAH61823-03
AAH61823-MD
2-Amino-3-bromobenzonitrile 95%
2-Amino-3-bromobenzonitrile
Purity: 95%
CAS No.: 114344-60-4
Molecular Formula: C7H5BrN2
Formula Weight: 197.03
Physical Form: Solid
UN No.: 3439
Formula:
C₇H₅BrN₂ Storage Temperature: Ambient |
MDL Number:
MFCD05670539 CAS Number: 114344-60-4 UN: 3439 ADR: 6.1,III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...