5-Fluoro-2-nitroaniline 97%

Supplier: Thermo Scientific Chemicals
Warning

H61806-06 H61806-14 H61806-22
AAH61806-06EA 48.66 CAD
AAH61806-06 AAH61806-14 AAH61806-22
5-Fluoro-2-nitroaniline 97%
5-Fluoro-2-nitroaniline

Purity: 97% CAS No.: 2369-11-1 Molecular Formula: C6H5FN2O2 Formula Weight: 156.11 Physical Form: Solid UN No.: 2811 MDL No.: MFCD00034065

Formula: C₆H₅FN₂O₂
MW: 156.12
Storage Temperature: Ambient
MDL Number: MFCD00034065
CAS Number: 2369-11-1
UN: 2811
ADR: 6.1 III

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR