potassium thiobenzoate 95%

Supplier: Thermo Scientific Chemicals
Warning

H61741-03 H61741-MD
AAH61741-03EA 303.36 CAD
AAH61741-03 AAH61741-MD
potassium thiobenzoate 95%
potassium thiobenzoate

Purity: 95% CAS No.: 28170-13-0 Molecular Formula: C7H5KOS Formula Weight: 176.28 Physical Form: Solid

Formula: C₇H₅KOS
Storage Temperature: Ambient
CAS Number: 28170-13-0

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR