2-Amino-3-fluorobenzonitrile 95%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61580-03
H61580-MD
AAH61580-03EA
214.88
CAD
AAH61580-03
AAH61580-MD
2-Amino-3-fluorobenzonitrile 95%
2-Amino-3-fluorobenzonitrile
Purity: 95%
CAS No.: 115661-37-5
Molecular Formula: C7H5FN2
Formula Weight: 136.13
Physical Form: Solid
UN No.: 3439
Formula:
C₇H₅FN₂ MW: 136.13 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD07368774 CAS Number: 115661-37-5 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...