3-Chloro-4-cyanobenzeneboronic acid 96%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: (3-Chloro-4-cyanophenyl)boronic acid

H61543-03 H61543-MD
AAH61543-03EA 862.68 CAD
AAH61543-03 AAH61543-MD
3-Chloro-4-cyanobenzeneboronic acid 96%
3-Chloro-4-cyanobenzeneboronic acid

Purity: 96% CAS No.: 1008415-02-8 Molecular Formula: C7H5BClNO2 Formula Weight: 164.93 Physical Form: Solid

Formula: C₇H₅BClNO₂
MW: 181.382 g/mol
Storage Temperature: Ambient
MDL Number: MFCD11504841
CAS Number: 1008415-02-8

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR