2-Cyano-4-(trifluoromethyl)phenylboronic acid 95%

Supplier: Thermo Scientific Chemicals
Warning

Synonyms: 2-Cyano-4-(trifluoromethyl)benzeneboronic acid

H61451-03 H61451-MD
AAH61451-03EA 676.24 CAD
AAH61451-03 AAH61451-MD
2-Cyano-4-(trifluoromethyl)phenylboronic acid 95%
2-Cyano-4-(trifluoromethyl)phenylboronic acid

Purity: 95% CAS No.: 1218790-84-1 Molecular Formula: C8H5BF3NO2 Formula Weight: 214.94 Physical Form: Solid

Formula: C₈H₅BF₃NO₂
MW: 214.94 g/mol
Storage Temperature: Ambient
MDL Number: MFCD13195679
CAS Number: 1218790-84-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR