Ethyl (3-fluorobenzoyl)acetate 98%
Supplier: Thermo Scientific Chemicals
H61315-03
H61315-06
H61315-MD
AAH61315-03EA
140.62
CAD
AAH61315-03
AAH61315-06
AAH61315-MD
Ethyl (3-fluorobenzoyl)acetate 98%
Ethyl (3-fluorobenzoyl)acetate
Purity: 98%
CAS No.: 33166-77-7
Molecular Formula: C11H11FO3
Formula Weight: 210.20
Physical Form: Liquid
Density: 1.166
Refractive Index: 1.5170
Appearance: Colorless to orange
Boiling Point: 262-263°
MDL No.: MFCD03424809
Formula:
C₁₁H₁₁FO₃ Boiling Pt: 262…263 °C Density: 1.166 Storage Temperature: Ambient |
MDL Number:
MFCD03424809 CAS Number: 33166-77-7 |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...