8-(Trifluoromethyl)quinoline 98%

Supplier: Thermo Scientific Chemicals
Warning

H61267-03 H61267-06
AAH61267-03EA 180.12 CAD
AAH61267-03 AAH61267-06
8-(Trifluoromethyl)quinoline 98%
8-(Trifluoromethyl)quinoline

Purity: 98% CAS No.: 317-57-7 Molecular Formula: C10H6F3N Formula Weight: 197.16 Physical Form: Solid

Formula: C₁₀H₆F₃N
MW: 197.16 g/mol
Storage Temperature: Ambient
MDL Number: MFCD09744668
CAS Number: 317-57-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR