2-Bromo-6-(4-morpholinyl)benzonitrile 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
H61193-03
H61193-MD
AAH61193-03EA
376.04
CAD
AAH61193-03
AAH61193-MD
2-Bromo-6-(4-morpholinyl)benzonitrile 98%
2-Bromo-6-(4-morpholinyl)benzonitrile
Purity: 98%
CAS No.: 1129540-65-3
Molecular Formula: C11H11BrN2O
Formula Weight: 267.13
Physical Form: Solid
UN No.: 3439
Formula:
C₁₁H₁₁BrN₂O MW: 267.126 g/mol Storage Temperature: Ambient |
CAS Number:
1129540-65-3 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...