1-(4-Cyanophenyl)-2,5-dimethylpyrrole 98%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
4-(2,5-Dimethyl-1-pyrrolyl)benzonitrile
H61178-03
H61178-06
AAH61178-03EA
111.71
CAD
AAH61178-03
AAH61178-06
1-(4-Cyanophenyl)-2,5-dimethylpyrrole 98%
1-(4-Cyanophenyl)-2,5-dimethylpyrrole
Purity: 98%
CAS No.: 119516-86-8
Molecular Formula: C13H12N2
Formula Weight: 196.25
Physical Form: Solid
UN No.: 3439
Formula:
C₁₃H₁₂N₂ MW: 196.25 g/mol Storage Temperature: Ambient |
MDL Number:
MFCD03228548 CAS Number: 119516-86-8 UN: 3439 ADR: 6.1 III |
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...