3-Bromo-5-(1-piperidinylcarbonyl)benzeneboronic acid 97%
Supplier: Thermo Scientific Chemicals
|
Warning
|
Synonyms:
Boronic acid, B-[3-bromo-5-(1-piperidinylcarbonyl)phenyl]-
H61113-03
H61113-MD
AAH61113-03EA
903.76
CAD
AAH61113-03
AAH61113-MD
3-Bromo-5-(1-piperidinylcarbonyl)benzeneboronic acid 97%
3-Bromo-5-(1-piperidinylcarbonyl)benzeneboronic acid
Purity: 97%
Molecular Formula: C12H15BBrNO3
Formula Weight: 311.97
Physical Form: Solid
Learn more

About VWR
Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...