6-Bromo-3,4-dihydro-2H-1,4-benzothiazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H61089-03 H61089-MD
AAH61089-03EA 350.76 CAD
AAH61089-03 AAH61089-MD
6-Bromo-3,4-dihydro-2H-1,4-benzothiazine 97%
6-Bromo-3,4-dihydro-2H-1,4-benzothiazine

Purity: 97% CAS No.: 187604-75-7 Molecular Formula: C8H8BrNS Formula Weight: 230.12 MDL No.: MFCD23379863

Formula: C₈H₈BrNS
MW: 230.123 g/mol
Storage Temperature: Ambient
MDL Number: MFCD23379863
CAS Number: 187604-75-7

Order Now


Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR