7-Nitro-3,4-dihydro-2H-1,4-benzothiazine 97%

Supplier: Thermo Scientific Chemicals
Warning

H61048-03 H61048-MD
AAH61048-03EA 410.8 CAD
AAH61048-03 AAH61048-MD
7-Nitro-3,4-dihydro-2H-1,4-benzothiazine 97%
7-Nitro-3,4-dihydro-2H-1,4-benzothiazine

Purity: 97% CAS No.: 69373-37-1 Molecular Formula: C8H8N2O2S Formula Weight: 196.22 MDL No.: MFCD23379852

Formula: C₈H₈N₂O₂S
Storage Temperature: Ambient
MDL Number: MFCD23379852
CAS Number: 69373-37-1

Order Now

Learn more

About VWR

Avantor is a vertically integrated, global supplier of discovery-to-delivery solutions for...

Learn more About VWR